Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ AEGIS KVS4™ Cut-Resistant Gloves

Specifications

Coating Material Sponge Nitrile
Color Black, Yellow
Cut Resistance Rating Level 4
Material Dupont™ Kevlar™ Aramid Fiber, Stainless Steel
For Use With (Application) Oily surfaces, Metal stamping, mechanical and automotive, sheet Metal assembly, Glass handling, bottling, recycling
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity & Availability  
19-120-645
View Documents
SHOWA™
25011
11 in. 27.9 cm 2X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-120-641
View Documents
SHOWA™
25007
7 in. 17.8 cm Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications

Specifications

Sponge Nitrile
Level 4
Oily surfaces, Metal stamping, mechanical and automotive, sheet Metal assembly, Glass handling, bottling, recycling
Black, Yellow
Dupont™ Kevlar™ Aramid Fiber, Stainless Steel
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit