Learn More
SHOWA™ AEGIS KVS4™ Gloves
Click to view available options
Size:
Medium
Large
X-Large
Specifications
Color | Black, Yellow |
Material | Nitrile, Aramid Fiber, Stainless Steel Fiber, Polyester |
Product Type | Cut-resistant Gloves |
Size | Medium |