Learn More
SHOWA Glove Polyurethane-Coated Delicate Operations Gloves
Supple coating provides good resistance to abrasion and micro-cuts
$24.50 - $24.75
Specifications
Cleaning Procedure | Launder at 40°C using a neutral detergent |
---|---|
Material | Polyurethane |
Glove Cuff Style | Knit |
Liner Material | Nylon |
Catalog Number | Mfr. No. | Coated Area | Color | Size | Exterior Finish | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Coated Area | Color | Size | Exterior Finish | Price | Quantity & Availability | ||
19-130-4682A
|
SHOWA
BO500WS |
Palm | White | Small | Textured |
Dozen of 12 for $24.75
Case of 6 DZ for $148.50 |
|
19-130-4682D
|
SHOWA
BO500WXL |
Palm | White | X-Large | Textured |
Dozen of 12 for $24.75
Case of 6 DZ for $148.50 |
|
19-130-4682B
|
SHOWA
BO500WM |
Palm | White | Medium | Textured |
Dozen of 12 for $24.75
Case of 6 DZ for $148.50 |
|
19-130-4682C
|
SHOWA
BO500WL |
Palm | White | Large | Textured |
Dozen of 12 for $24.75
Case of 6 DZ for $148.50 |
|
19-130-4683C
|
SHOWA
BO500BL |
Palm | Black | Large | Textured |
Dozen of 12 for $24.75
Case of 6 DZ for $148.50 |
|
19-130-4683D
|
SHOWA
BO500BXL |
Palm | Black | X-Large | Textured |
Dozen of 12 for $24.75
Case of 6 DZ for $148.50 |
|
19-130-4683A
|
SHOWA
BO500BS |
Palm | Black | Small | Textured |
Dozen of 12 for $24.75
Case of 6 DZ for $148.50 |
|
19-130-4683B
|
SHOWA
BO500BM |
Palm | Black | Medium | Textured |
Dozen of 12 for $24.75
Case of 6 DZ for $148.50 |
|
19-130-4684D
|
SHOWA
BO600XL |
Fingertip | White | X-Large | Smooth |
Dozen of 12 for $24.50
Case of 6 DZ for $147.00 |
|
19-130-4684C
|
SHOWA
BO600L |
Fingertip | White | Large | Smooth |
Dozen of 12 for $24.50
Case of 6 DZ for $147.00 |
|
19-130-4684B
|
SHOWA
BO600M |
Fingertip | White | Medium | Smooth |
Dozen of 12 for $24.50
Case of 6 DZ for $147.00 |
|
19-130-4684A
|
SHOWA
B0600S |
Fingertip | White | Small | Smooth |
Dozen of 12 for $24.50
Case of 6 DZ for $147.00 |
|
Description
The coating on “palm fit” style is carefully demarcated to follow the shape of the palm and effectively protects the contact surface while leaving the back of the hand ventilated to limit perspiration. Coating on “top fit” style is confined to the fingertips and allows small parts to be handled with ease while protecting them from fingerprints.
- Breathable microporous polyurethane coating guarantees an unequaled level of comfort for prolonged use
- 13 gauge seamless nylon liner offers flexibility and precision while eliminating the risk of irritation
- Anatomical shape reduces hand fatigue
- Elasticated cuff keeps glove correctly adjusted
Order Info
Launderable (at 40°C using a neutral detergent).
Specifications
Launder at 40°C using a neutral detergent | |
Polyurethane | |
Knit | |
Nylon |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.