Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ N-DEX™ 6005PF Disposable Nitrile Gloves

N-DEX™ 6005 series powder free gloves provide enhanced dexterity and performance across a range of tactical applications including food, electronics, janitorial, municipal services and utilities.

$413.33

Specifications

Certifications/Compliance 21 CFR 170-199
Applications Industrial
AQL 1.5
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-120-2415A
View Documents
SHOWA™
6005PFXS
X-Small
Case of 2000 for $413.33
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-120-2415B
View Documents
SHOWA™
6005PFSM
Small
Case of 2000 for $413.33
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-120-2415C
View Documents
SHOWA™
6005PFMD
Medium
Case of 2000 for $413.33
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-120-2415D
View Documents
SHOWA™
6005PFL
Large
Case of 2000 for $413.33
Add to cart
 
19-120-2415E
View Documents
SHOWA™
6005PFXL
X-Large
Case of 2000 for $413.33
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

N-DEX™ 6005 series powder free gloves provide enhanced dexterity and performance across a range of tactical applications including food, electronics, janitorial, municipal services and utilities.

  • Easy to put on and remove
  • Can easily be one-hand used
  • Ambidextrous
  • Rolled cuff
  • Smooth grip
  • Made in the U.S.A.
  • Specific applications include food handling, agriculture, washing and cleaning, public utility and coroner/morgue
Specifications

Specifications

21 CFR 170-199
1.5
0.10 mm
9.5 in.
Industrial
4 mil
No Texture
24 cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit