Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Neo Grab™ Neoprene Gloves

Offer superior durability and chemical protection

$160.60 - $299.50

Specifications

Coating Material Rough, wrinkle
Color Black
Material Neoprene
Size 10
For Use With (Application) manufacturing, refining, Petrochemicals, degreasing, plating, handling Oils, Acids, Caustics, Alcohols and Solvents
View More Specs

Products 3
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
11-395-22
View Documents
SHOWA™
6797R10
Elbow length 30.48 cm Dozen of 12 for $299.50
Case of 3 DZ for $759.00
Add to cart
 
11-395-9
View Documents
SHOWA™
6780R10
12 in. 30.48 cm Dozen of 12 for $160.60
Case of 6 DZ for $812.50
Add to cart
 
19-019202
View Documents
SHOWA™
6784R10
14 in. 35.56 cm Dozen of 12 for $230.00
Case of 3 DZ for $584.31
Add to cart
 
Description

Description

Provide abrasion and cut resistance.

  • Multidipped neoprene gauntlet gloves offer excellent chemical protection
  • Heavyweight gloves feature wrinkle finish for better wet gripping
  • May be worn over unsupported chemical resistant gloves for added protection
  • With Sanitized™ cotton liner

Specifications

Specifications

Rough, wrinkle
Neoprene
manufacturing, refining, Petrochemicals, degreasing, plating, handling Oils, Acids, Caustics, Alcohols and Solvents
Sanitized Cotton
Black
10
Gauntlet
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit