SHOWA™ Nitri-Pro™ Gloves

Ideal replacements for heavy cotton or leather work gloves

Manufacturer: SHOWA™ 7000PR09

 View more versions of this product

Catalog No. 19-014669

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Launder at 40°C using a neutral Detergent
    Exterior Finish Rough
    Glove Cuff Style Knit
    For Use With (Application) heavy duty handling of castings, cores and fabricated Metals, brick, tile, plywood, drywall
    Material Nitrile
    Liner Material Cotton Jersey
    Size 9
    Coated Area Palm
    Color White, navy
    Cut Resistance Rating Level 2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit