Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA T-Flex Cut-Resistant Gloves
Thin, lightweight glove boasts a construction made up of only Dyneema cut-resistant fibers
Specifications
Color | Light Gray |
---|---|
Material | AlphaSan™, Dyneema™ |
For Use With (Application) | meat cutting, construction, glove liner, recycling, bottling, Glass handling, sheet Metal handling, razor blades |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-130-1055
|
SHOWA
811507 |
Small |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-130-1056
|
SHOWA
811508 |
Medium |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-130-1057
|
SHOWA
811509 |
Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-130-1054
|
SHOWA
811510 |
X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-169-950
|
SHOWA
811506 |
X-Small |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Outstanding ratio of cut resistance to weight
- Good touch and tactile sensitivity
- Suits assembly workers handling jagged rivets and sharp parts, as well as machinists replacing pinions and sharp-edged tooling parts
- Ambidextrous
- Thickness: 15-gauge
- Made in the U.S.A.
Compliance
Meet FDA CR21 requirements as an under-glove for meat cutting
Specifications
Light Gray | |
AlphaSan™, Dyneema™ | |
meat cutting, construction, glove liner, recycling, bottling, Glass handling, sheet Metal handling, razor blades |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title