SHOWA™ Zorb-IT™ Work Gloves

Conform and adhere to seamless nylon shell

Manufacturer: SHOWA™ 455009

 View more versions of this product

Catalog No. 19-130-1301D

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Sponge
    Glove Cuff Style Knit
    For Use With (Application) material handling, Metal fabrication, durable goods assembly
    Liner Material Nylon
    Size 9
    Cleaning Procedure Launder at 40°C using a neutral Detergent
    Material Nitrile
    Coated Area Palm
    Color Gray, white

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit