Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120677

This item is not returnable.
View return policy
Description
Positive test controls include: 293T, BxPC-3, Mouse brain, Rat heart. The target is usually found in the following locations: Nucleus.
SHOX Polyclonal antibody specifically detects SHOX in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
SHOX | |
Polyclonal | |
Unconjugated | |
SHOX | |
GCFX; growth control factor, X-linked; PHOG; Pseudoautosomal homeobox-containing osteogenic protein; short stature homeobox; short stature homeobox protein; Short stature homeobox-containing protein; SHOX; SHOXY; SS | |
A synthetic Peptide of human SHOX | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
0.701 mg/mL | |
PBS with 0.02% sodium azide; pH 7.3 | |
O15266 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6473 | |
4° C, do not freeze | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title