Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHP-1 Mouse anti-Human, Clone: CPTC-PTPN6-2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30715810UG

 View more versions of this product

Catalog No. NB121789


Add to cart

Description

Description

SHP-1 Monoclonal antibody specifically detects SHP-1 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

SHP-1
CPTC-PTPN6-2
Unconjugated
10 mM PBS with 0.05% BSA
HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1
Recombinant human full-length SHP-1 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Western Blot, ELISA
0.2 mg/ml
Western Blot 1-2 ug/ml, ELISA
Purified
Mouse
IgG2a Kappa
100 μg
Apoptosis, Phospho Specific, Protein Phosphatase
Primary
5777
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit