Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP-2/PTPN11 Rabbit anti-Human, Mouse, Clone: SN07-32, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267715
Description
SHP-2/PTPN11 Monoclonal antibody specifically detects SHP-2/PTPN11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin).Specifications
SHP-2/PTPN11 | |
SN07-32 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
Rabbit | |
IgG | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200 | |
Purified | |
PTPN11 | |
Recombinant Protein | |
Protein A purified | |
Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
Primary | |
5781 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title