Learn More
Medchemexpress LLC SHP099 monohydrochloride | 2200214-93-1 | 99.9% | 388.72 | C16H20Cl3N5 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY100388A500MG
Encompass_Preferred
SHP099 monohydrochloride is the hydrochloride salt of a potent, selective, allosteric SHP2 inhibitor used to probe SHP2-dependent signaling in cellular and preclinical studies. It stabilizes SHP2 in an autoinhibited conformation, suppresses RAS-ERK signaling, and has demonstrated efficacy in models driven by receptor tyrosine kinases.
  • Potent SHP2 inhibition (IC50 ≈ 70 nM).
  • High purity suitable for biological studies.
  • Orally bioavailable in preclinical models.
  • Useful for in vitro and in vivo signaling pathway studies.
  • Available in multiple pack sizes for flexible experimental needs.
Catalog No. 50-002-12739