Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHPRH Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550788

Catalog No. PA550788


Add to cart

Description

Description

The antibody detects endogenous levels of total SHPRH protein.

SHPRH is a ubiquitously expressed protein that contains motifs characteristics of several DNA repair proteins, transcription factors, and helicases. SHPRH is a functional homolog of S. cerevisiae RAD5 (Unk et al., 2006 [PubMed 17108083]).
Specifications

Specifications

SHPRH
Polyclonal
Unconjugated
SHPRH
Liquid
SHPRH
Fusion protein corresponding to residues near the C terminal of human SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.4 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q149N8
KIAA2023
Rabbit
IgG
100 μL
-20°C
Primary
257218
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit