Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SHQ1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570281
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%.
SHQ1 is the human homolog of yeast Shq1p, a protein that acts as an accessory factor in the assembly of H/ACA ribonucleoproteins. SHQ1 has been shown to bind the major H/ACA core protein and the pseudouridine synthase NAP57, but precedes the assembly role of NAF1 and of the other H/ACA core proteins. SHQ1 assists in the assembly of H/ACA-box ribonucleoproteins that function in the processing of ribosomal RNAs, modification of spliceosomal small nuclear RNAs, and stabilization of telomerase.
Specifications
SHQ1 | |
Polyclonal | |
Unconjugated | |
SHQ1 | |
2810403P18Rik; FLJ10539; gene associated with retinoid and interferon-induced mortality 1; GRIM-1; hypothetical protein LOC507486; protein SHQ1 homolog; RGD1310610; Shq1; SHQ1 homolog; SHQ1 homolog (S. cerevisiae); SHQ1, H/ACA ribonucleoprotein assembly factor; Shq1p; zgc:158255 | |
Rabbit | |
Affinity chromatography | |
RUO | |
72171 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7TMX5 | |
SHQ1 | |
synthetic peptide directed towards the N-terminal region of mouse SHQ1. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction