Learn More
Invitrogen™ SI-CLP Monoclonal Antibody (1C11)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA549028
Description
In vivo, the monoclonal antibody 1C11 recognizes SI-CLP in the cellular fraction of bronchoalveolar lavage specimens obtained from patients with chronic inflammatory disorders of the respiratory tract and in peripheral-blood leukocytes (PBLs) from these patients as well as healthy donors.
SI-CLP detects human Stabilin-interacting chitinase like protein (SI-CLP), a novel member of the Glyco-18-domain containing protein family. SI-CLP is a secretory protein which is up-regulated in response to interleukin-4 and dexamethasone in activated macrophages. SI-CLP is also expressed by T- and B-lymphocytes, and by cells of monocytic and epithelial origin. SI-CLP is the ligand for stabilin-1, an intracellular sorting receptor which sorts SI-CLP into late endosomes and secretory lysosomes in activated macrophages.
Specifications
SI-CLP | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
Q9BWS9 | |
CHID1 | |
Amino acids 24 to 73 fragment GST-fusion protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry | |
1C11 | |
Unconjugated | |
CHID1 | |
3110023E09Rik; AI841869; CHID1; chitinase domain containing 1; chitinase domain-containing protein 1; FLJ42707; GL008; MGC3234; PSEC0104; SB139; SICLP; SI-CLP; stabilin-1 interacting chitinase-like protein; stabilin-1-interacting chitinase-like protein | |
Rat | |
Protein G | |
RUO | |
66005 | |
4°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.