Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SI Manufacturing Compact Digital Scale with Bowl Products for Science Education

For weighing multiple small objects at once.

Manufacturer:  SI Manufacturing 39750

Catalog No. S04758


Add to cart

Includes: Clear bowl

Description

Description

  • Comes with large display screen
  • 2kg weight

Specifications

Specifications

2 kg
Digital Compact Scale
Clear bowl
4.4 lb.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit