Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sialin/SLC17A5 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | Sialin/SLC17A5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB024035
|
Novus Biologicals
NBP294758100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB024036
|
Novus Biologicals
NBP29475820UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Sialin/SLC17A5 Polyclonal antibody specifically detects Sialin/SLC17A5 in Mouse, Rat samples. It is validated for Western BlotSpecifications
Sialin/SLC17A5 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
26503 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
ASTSodium/sialic acid cotransporter, FLJ22227, FLJ23268, ISSD, Membrane glycoprotein HP59, NSD, SD, sialic acid storage disease, SIALIN, SIASD, SLDSolute carrier family 17 member 5, solute carrier family 17 (anion/sugar transporter), member 5, solute carrier family 17, member 5 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC17A5 (NP_036566.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title