Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIB 1553A hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
191611-89-9
C13H20ClNOS
273.819
RJSVKPQKXSMZMT-UHFFFAOYSA-N
sib 1553a hydrochloride,sib 1553a hyrdrochloride,d00eco,+--4-2-1-methyl-2-pyrrolidinyl ethyl thio phenol hydrochloride,+/--4-2-1-methyl-2-pyrrolidinyl ethyl thio phenol hydrochloride,4-2-1-methylpyrrolidin-2-yl ethyl sulfanyl phenol hydrochloride,phenol, 4-2-1-methyl-2-pyrrolidinyl ethyl thio-, hydrochloride
9881989
4-[2-(1-methylpyrrolidin-2-yl)ethylsulfanyl]phenol;hydrochloride
CN1CCCC1CCSC2=CC=C(C=C2)O.Cl
This item is not returnable. View return policy
191611-89-9
C13H20ClNOS
273.819
RJSVKPQKXSMZMT-UHFFFAOYSA-N
sib 1553a hydrochloride,sib 1553a hyrdrochloride,d00eco,+--4-2-1-methyl-2-pyrrolidinyl ethyl thio phenol hydrochloride,+/--4-2-1-methyl-2-pyrrolidinyl ethyl thio phenol hydrochloride,4-2-1-methylpyrrolidin-2-yl ethyl sulfanyl phenol hydrochloride,phenol, 4-2-1-methyl-2-pyrrolidinyl ethyl thio-, hydrochloride
9881989
4-[2-(1-methylpyrrolidin-2-yl)ethylsulfanyl]phenol;hydrochloride
CN1CCCC1CCSC2=CC=C(C=C2)O.Cl

Subunit selective nAChR agonist

Nicotinic acetylcholine receptor (nAChR) agonist. Displays selectivity for β4 subunit-containing receptors. Stimulates acetylcholine levels and other neurotransmitters relevant for cognitive processes. Improves attention deficits and memory performance in a Parkinson's disease model.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 273.82
Percent Purity >98%
Chemical Name or Material SIB 1553A hydrochloride

Recommended Storage : Store at +4°C