Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SID 26681509
Cathepsin L inhibitor
$339.88 - $1424.48
Specifications
Purity | >97% |
---|---|
Product Type | SID 26681509 |
Molecular Weight (g/mol) | 539.65 |
Inhibitors | SID 26681509 |
Description
Potent and reversible human cathepsin L inhibitor (IC50 = 56 nM). Displays no inhibitory activity at cathepsin G. Antimalarial; inhibits leishmaniasis.Specifications
>97% | |
SID 26681509 | |
539.65 | |
SID 26681509 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title