Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Side-Mount Acrylic Glove Box Holders

Convenient side-mount design

$48.00 - $72.50

Specifications

Material Acrylic
Mounting Horizontal or vertical
Product Type Glove Dispenser
Depth (English) 4 in.
Depth (Metric) 10.16cm
View More Specs

Products 9
Catalog Number Mfr. No. Dimensions (D x W x H) Interior Style Holds Color Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (D x W x H) Interior Style Holds Color Price Quantity & Availability  
06-666-819
View Documents
Mitchell Plastics
MGSM1000
10.16 x 25.4 x 13.9cm (4 x 10 x 5.5 in.) Side mount; Single Glove Box Holder 1 box Clear
Each for $48.00
Add to cart
 
06-666-822
View Documents
Mitchell Plastics
MGSM2000
10.16 x 50.8 x 13.9cm (4 x 20 x 5.5 in.) Side mount; Double Glove Box Holder 2 boxes Clear
Each for $59.00
Add to cart
 
06-666-825
View Documents
Mitchell Plastics
MGSM3000
10.16 x 76.2 x 13.9cm (4 x 30 x 5.5 in.) Side mount Clear
Each for $64.50
Add to cart
 
06-666-827
View Documents
Mitchell Plastics
MGSM3001R
10.16 x 76.2 x 13.9cm (4 x 30 x 5.5 in.) Side mount 3 Boxes Neon Red
Each for $72.50
Add to cart
 
06-666-820
View Documents
Mitchell Plastics
MGSM1001G
10.16 x 25.4 x 13.9cm (4 x 10 x 5.5 in.) Side mount; Single Glove Box Holder 1 box Neon green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
06-666-821
View Documents
Mitchell Plastics
MGSM1001R
10.16 x 25.4 x 13.9cm (4 x 10 x 5.5 in.) Side mount; Single Glove Box Holder 1 box Neon Red This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
06-666-824
View Documents
Mitchell Plastics
MGSM2001R
10.16 x 50.8 x 13.9cm (4 x 20 x 5.5 in.) Side mount Neon Red This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
06-666-823
View Documents
Mitchell Plastics
MGSM2001G
10.16 x 50.8 x 13.9cm (4 x 20 x 5.5 in.) Side mount; Double Glove Box Holder 2 boxes Neon Green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
06-666-826
View Documents
Mitchell Plastics
MGSM3001G
10.16 x 76.2 x 13.9cm (4 x 30 x 5.5 in.) Side mount 3 Boxes Neon green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • May be mounted horizontally or vertically
  • Two holes and double-sided foam tape are provided for wall mounting
Specifications

Specifications

Acrylic
Glove Dispenser
10.16cm
5.5 in.
Horizontal or vertical
4 in.
Horizontal design for use under cabinets, shelves and tight spaces
13.9cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit