Learn More
Chem-Impex International, Inc.Supplier Diversity Partner 9-Fmoc-aminoxanthen-3-yloxy polystyrene resin, , CAS#: 915706-90-0, 5G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 035415G
9-Fmoc-aminoxanthen-3-yloxy polystyrene resin, , CAS#: 915706-90-0, 5G. Hazardous Material: NO. MDL #: MFCD28053610. Grain Size: 100-200 mesh (Microscope). Substitution: 0.6-0.8 meq/g (UV spectroscopy after coupling with Fmoc-leu-OH). DVB Percentage: 1% DVB. Appearance: Pale white or slight yellow beads. Storage Temperature: Store at 0-8 °C. Additional Comments: Excellent resin for the synthesis of protected peptide amides and carboxamides. Cleavage is effected by 1% TFA.
Catalog No. 50-498-003