Siemens Healthineers Hema-Chek™ Fecal Occult Blood Test Slides and Developer
This simple, economical test produces results in 30 seconds.
Manufacturer: Siemens Healthineers 2596
 View more versions of this product
Catalog No. 23-042-352
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Developer
    For Use With (Application) Detection of fecal occult Blood
    Quantity 10 Case
    Test Time 30 sec.
  • Description & Specifications
Related Products
Product Content Feedback