Learn More
W.S. TYLER™ Sieve Certification and Recertification Services
Certification services offered for new or used sieves
Supplier: W.S. TYLER™ 9998FRC
Description
Certification Service
- Order when purchasing new sieves
- Uses a computerized video imaging system to verify that each sieve opening meets ASTM™ and ISO specifications within a tolerance of ±0.1 microns
- Sieves are then serialized and supplied with documentation, traceable to NIST™, detailing the openings and the wire diameters of the mesh
- Offer the same examination and documentation processes for in-use sieves as for new sieves
- Contact your Fisher Scientific Customer Service Representative for the recommended sieve aperture/recertification schedule
Specifications
Recertification Service for used sieves | |
To serialize and supply with documentation |