Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SIGIRR Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
$172.84 - $532.44
Specifications
Antigen | SIGIRR |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
SIGIRR Polyclonal specifically detects SIGIRR in Mouse samples. It is validated for Western Blot.Specifications
SIGIRR | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
IL-1R8, single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8, TIR8MGC110992, Toll/interleukin-1 receptor 8 | |
SIGIRR | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9JLZ8 | |
59307 | |
Mouse myeloma cell line NS0-derived recombinant mouse SIGIRR Met1-His117 Accession # Q9JLZ8 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse SIGIRR in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) SIGIRR, rhIL-1 R9, recombinant mouse (rm) IL-18 R beta , rmIL-18 R, and recombinant rat IL-1 R6 is observed. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title