Learn More
Siglec-1/CD169 Mouse anti-Human, Alexa Fluor™ 750, Clone: 7-239, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP237729AF750
Catalog No. NP237729A70
Add to cart
Description
Description
Siglec-1/CD169 Monoclonal antibody specifically detects Siglec-1/CD169 in Human samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen).
Specifications
Specifications
Siglec-1/CD169
7-239
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody 7-239 recognizes CD169 (sialoadhesin, Siglec-1), a 210 kDa type I transmembrane glycoprotein expressed on macrophages and dendritic cells.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
SIGLEC1
Human rhinovirus 14-infected monocyte-derived dendritic cells.
0.1 mL
Primary
6614
Human