Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-1/CD169 Rat anti-Mouse, DyLight 550, Clone: RM0531-7E46, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241751R

Catalog No. NBP241751R

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Siglec-1/CD169
RM0531-7E46
Western Blot, Flow Cytometry
Purified
SIGLEC1
Recombinant Mouse Siglec-1; aa20-aa639.
Protein G purified
Adaptive Immunity, Immunology
Primary
6614
Western Blot, Flow Cytometry
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit