Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-15 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP2411620.025MG
Description
Siglec-15 Polyclonal specifically detects Siglec-15 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Siglec-15 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
SIGLEC15 | |
Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human SIGLEC15. The immunogen is located within amino acids 50 - 100 of SIGLEC15. Amino Acid Squence: TFTHPHRHYDGPLT | |
Affinity Purified | |
RUO | |
284266 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 - 2 μg/mL, Flow Cytometry, ELISA, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
CD33 antigen-like 3SIGLEC-15, CD33 molecule-like 3, CD33L3, HsT1361, sialic acid binding Ig-like lectin 15, sialic acid-binding Ig-like lectin 15, Siglec-15 | |
Rabbit | |
42 kDa | |
0.025 mg | |
Primary | |
SIGLEC15 antibody is predicted to not cross-react with other SIGLEC family members. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction