Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-15 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241162DL594
Description
Siglec-15 Polyclonal specifically detects Siglec-15 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Siglec-15 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
284266 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
CD33 antigen-like 3SIGLEC-15, CD33 molecule-like 3, CD33L3, HsT1361, sialic acid binding Ig-like lectin 15, sialic acid-binding Ig-like lectin 15, Siglec-15 | |
Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human SIGLEC15. The immunogen is located within amino acids 50 - 100 of SIGLEC15. Amino Acid Squence: TFTHPHRHYDGPLT | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction