Learn More
Siglec-3/CD33 Mouse anti-Human, Alexa Fluor™ 532, Clone: 44M12D3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222377AF532
Catalog No. NP222377A32
Add to cart
Description
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Siglec-3/CD33
44M12D3
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD33
Amino acids 42-159 of human CD33 were used as the immunogen for the antibody.
0.1 mL
Primary
945