Learn More
Siglec-3/CD33 Mouse anti-Human, Alexa Fluor™ 594, Clone: 996816, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB11372T100UG
Catalog No. FB11372T100
Add to cart
This item is not returnable. View return policy
Description
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Siglec-3/CD33
996816
Flow Cytometry 0.25-1 μg/106 cells
P20138
CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67
Mouse myeloma cell line, NS0-derived human Siglec-3, Asp18-His259, Accession # P20138
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Siglec-3 in direct ELISAs.
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
945
Human