Learn More
Siglec-3/CD33 Mouse anti-Human, HRP, Clone: 2C6B7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237388H
Catalog No. NBP237388H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Siglec-3/CD33
2C6B7
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CD33
Purified recombinant fragment of human CD33 (AA: 15-237) expressed in E. Coli.
Protein A or G purified
Immunology
Primary
945
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
HRP
PBS with No Preservative
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human