Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Siglec-3/CD33, Rabbit anti-Human, PE-Cy7, Clone: Gemtuzumab, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP275906PECY7100UL
Catalog No. NB009312
Description
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human Samples. It is validated for Flow Cytometry.
Specifications
Specifications
Siglec-3/CD33
Gemtuzumab
Flow Cytometry
Purified
CD33
CD33 is a transmembrane receptor expressed on predominantly on cells of the myeloid lineage, but it can also be found on some lymphoid cells.
Protein A purified
RUO
Store at 4°C in the dark. Do not freeze.
Monoclonal
Recognises human CD33.
Flow Cytometry
PE-Cyanine7
PBS
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Rabbit
IgG Kappa
100 uL
Immunology
Primary
945
Human