Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Siglec-5/CD170 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793695PLATES

 View more versions of this product

Catalog No. NB009778


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human Siglec-5/CD170.
Specifications

Specifications

234.38 pg/mL
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human Siglec-5/CD170 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of mouse anti-Human Siglec-5/CD170 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 685 ng of recombinant Human Siglec-5/CD170.
  • RUO
    This solid phase sandwich ELISA utilizes an antibody specific for Human Siglec-5/CD170.
    Siglec-5/CD170
    5 Plates
    Sandwich ELISA
    CD170, CD170 antigen, CD33 antigen-like 2, CD33L2OB binding protein-2, OB-BP2OB-binding protein 2, OBBP2sialic acid-binding Ig-like lectin 5, Obesity-binding protein 2, sialic acid binding Ig-like lectin 5, sialic acid-binding immunoglobulin-like lectin 5, SIGLEC-5
    HRP
    8778
    Antibody Pairs
    234.38-15000 pg/mL
    Storage is content dependent.
    Rabbit/Mouse
    Human
    SIGLEC5
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 15000 pg/mL is recommended.
    SDS
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit