Learn More
Siglec-7/CD328 Mouse anti-Human, Alexa Fluor™ 594, Clone: 6-434, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP237732AF594
Catalog No. NP237732A94
Description
Description
Siglec-7/CD328 Monoclonal antibody specifically detects Siglec-7/CD328 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Siglec-7/CD328
6-434
Flow Cytometry
Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec-7
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody 6-434 recognizes CD328 (Siglec-7), a 75 kDa transmembrane glycoprotein expressed mainly on NK cells, dendritic cells and monocytes. HLDA 8; WS Code ARN-80652
Flow Cytometry
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
SIGLEC7
Human dendritic cells.
0.1 mL
Primary
27036
Human