Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-9 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0552-6K12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211905AF350
Description
Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Western Blot.Specifications
Siglec-9 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
SIGLEC9 | |
Human recombinant Siglec-9 (extracellular domain) | |
Cardiovascular Biology | |
27180 | |
Store at 4°C in the dark. |
Western Blot | |
MM0552-6K12 | |
Western Blot | |
CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9 | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title