Learn More
Siglec-9 Mouse anti-Human, Alexa Fluor 488, Clone: MM0552-6K12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211905AF488
Catalog No. NB211905488
Add to cart
Description
Description
Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Siglec-9
MM0552-6K12
Western Blot
Purified
SIGLEC9
Extracellular domain of recombinant human Siglec-9.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9
Mouse
IgG2
0.1 ml
Primary
27180