Learn More
Siglec-F Rat anti-Mouse, BV421, Clone: E50-2440, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 565934
Catalog No. BDB565934
Add to cart
Description
Description
Siglec-F Monoclonal antibody specifically detects Siglec-F in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
Siglec-F
E50-2440
Brilliant Violet 421
Purified
Rat
Rat LOU, also known as Louvain, LOU/C, LOU/M IgG2a, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Siglec5; sialic acid binding Ig-like lectin 5; SIGL5; Siglec-5; CD170
Mouse Siglec-F and human IgG Fc recombinant fusion protein
25μg
Cell Biology
Primary
Murine
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.