Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-G Mouse anti-Mouse, APC, Clone: SH2.1, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 17583380

 View more versions of this product

Catalog No. 50-112-3146


Add to cart

Description

Description

Description: The SH2.1 monoclonal antibody reacts with mouse siglec-G, which is also known as sialic acid-binding immunoglobulin-like lectin-G. Siglec-G is an inhibitory receptor that is expressed in a B cell-specific manner with B1 B cells expressing the highest levels of siglec-G. When overexpressed, siglec-G inhibits B cell receptor-mediated calcium signaling. Siglec-G-deficient mice show a massive expansion of the B1a cell population as well as higher titers of natural IgM antibodies. Siglec-G-deficient B1a cells show a lower level of spontaneous apoptosis and a prolonged life span, which may result from higher expression levels of the transcription factor NFATc1. Siglec-G-dependent negative regulation exists in B1 cells, which may explain the naturally muted signaling response of B1 cells. Applications Reported: This SH2.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SH2.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.

2 µm post-manufacturing filtered. Siglec-G is an inhibitory receptor that is expressed in a B cell-specific manner with B1 B cells expressing the highest levels of siglec-G. When overexpressed, siglec-G inhibits B cell receptor-mediated calcium signaling. Siglec-G-deficient mice show a massive expansion of the B1a cell population as well as higher titers of natural IgM antibodies. Siglec-G-deficient B1a cells show a lower level of spontaneous apoptosis and a prolonged life span, which may result from higher expression levels of the transcription factor NFATc1. Siglec-G-dependent negative regulation exists in B1 cells, which may explain the naturally muted signaling response of B1 cells.
Specifications

Specifications

Siglec-G
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Liquid
Siglecg
IgG1, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
243958
Flow Cytometry
SH2.1
APC
Siglecg
Sialic Acid-Binding Immunoglobulin-Like Lectin-G; SiglecG; Siglec10; Siglec 10
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.