Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-H Rat anti-Mouse, Alexa Fluor™ 532, Clone: 23M15C8, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227062AF532

Catalog No. NP227062A32


Add to cart

Description

Description

Siglec-H Monoclonal antibody specifically detects Siglec-H in Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications

Specifications

Siglec-H
23M15C8
Immunohistochemistry, Immunohistochemistry-Paraffin
6430529G09Rik, Siglec-H, Siglech sialic acid binding Ig-like lectin H
Rat
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Siglech
A portion of amino acids 50-100 of mouse Siglec-H was used as the immunogen for this antibody.
0.05 mL
Primary
233274
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit