Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIGLEC10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555501
Description
Immunogen sequence: DGRFWIRVQE SVMVPEGLCI SVPCSFSYPR QDWTGSTPAY GYWFKAVTET TKGAPVATNH QSREVEMSTR GRFQLTGDPA KGNCSLVIRD AQMQDESQY Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 47%.
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6.
Specifications
SIGLEC10 | |
Polyclonal | |
Unconjugated | |
SIGLEC10 | |
PRO940; sialic acid binding Ig like lectin 10; sialic acid binding Ig-like lectin 10; sialic acid binding Ig-like lectin 10 Ig-like lectin 7; sialic acid binding Ig-like lectin G; sialic acid-binding Ig-like lectin 10; Siglec; SIGLEC10; Siglec-10; Siglecg; siglec-like gene 2; siglec-like protein 2; SLG2; UNQ477/PRO940 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
89790 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LC7 | |
SIGLEC10 | |
Recombinant protein corresponding to Human SIGLEC10. Recombinant protein control fragment (Product #RP-90295). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction