Learn More
ACROBiosystems Recombinant Protein;Human Siglec-5 / CD170 Protein, Fc Tag;HEK293;100UG;SI5,SIGLEC5,OB-BP2,CD170,CD33L2,OBBP2,SIGLEC-5,CD33-L2
Supplier:  ACROBiosystems SI5H5250100UG
Encompass_Preferred
Recombinant Protein;Human Siglec-5 / CD170 Protein, Fc Tag;100UG;Human Siglec-5, Fc Tag (SI5-H5250) is expressed from human 293 cells (HEK293). It contains AA Glu 17 - Thr 434 (Accession # AAH29896).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8544
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.