Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Hematoxylin Gill Solution

Nuclear stains intended for use in histology and cytology

$376.50 - $837.50

Specifications

For Use With (Application) Used with hematoxylin and eosin staining
Color Dark Reddish-Purple
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
SDGHS1128
Accessories View Documents
Sigma Diagnostics™
GHS11284L
4L Gill No. 1 Hematoxylin Solution
Each for $837.50
Add to cart
 
SDGHS280
Accessories View Documents
Sigma Diagnostics™
GHS2802.5L
2.5L Gill No. 2 Hematoxylin Solution
Each for $837.50
Add to cart
 
SDGHS332
Accessories View Documents
Sigma Diagnostics™
GHS3321L
1L Gill No. 3 Hematoxylin Solution
Each for $376.50
Add to cart
 
SDGHS380
Accessories View Documents
Sigma Diagnostics™
GHS3802.5L
2.5L Gill No. 3 Hematoxylin Solution
Each for $656.50
Add to cart
 
Description

Description

Generally, hematoxylins are classified as progressive or regressive based on dye concentration. Progressive stains have a lower concentration of dye and selectively stain nuclear chromatin. The desired intensity is a function of time. Regressive stains color all nuclear and cytoplasmic structures intensely. To arrive at correct chromatic response, excess dye must be removed by treatment with dilute acid (differentiation)

  • Gill No. 1 formulation is used as a progressive cytology stain
  • Gill No. 2 and No. 3 formulations may be used as progressive or regressive stains depending on length of staining time

For in vitro diagnostic use

Specifications

Specifications

Used with hematoxylin and eosin staining
Dark Reddish-Purple
Safety and Handling

Safety and Handling

ShelfLife : Expiry date on the label

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit