Learn More
Sigma Diagnostics™ Trichrome Stain Solution
Used primarily for distinguishing collagen from muscle tissue
$968.00 - $981.00
Specifications
Color Dark Purple
View More Specs
Products 2
Catalog Number Mfr. No. Product Type For Use With (Application) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type For Use With (Application) Includes Price Quantity & Availability  
SDHT10316
Accessories View Documents
Sigma Diagnostics™
HT10316500ML
Trichrome Stain LG Solution Study of connective tissue, muscle and collagen Fibers, stains collagen green, in vitro diagnostic use Chromotrope 2R 0.6%, Fast Green FCF (certified) 0.1%, Phosphotungstic Acid 0.8% and Acetic Acid 1%, Reagent stains collagen green
Each for $968.00
Add to cart
SDHT10516
Accessories View Documents
Sigma Diagnostics™
HT10516500ML
Trichrome Stain AB Solution Study of connective tissue, muscle and collagen Fibers, stains collagen blue and muscle red, in vitro diagnostic use Chromotrope 2R 0.6%, Aniline blue (certified) 0.3%, Phosphotungstic Acid 0.8% and Acetic Acid 1%, Reagent stains collagen blue
Each for $981.00
Add to cart
Description
Description
  • Trichrome Stain LG Solution contains Chromotrope 2R, 0.6%, fast green FCF, (certified), 0.1%, phosphotungstic acid, 0.8% and acetic acid, 1.0%. Reagent stains collagen green
  • Trichrome Stain AB Solution contains Chromotrope 2R, 0.6%, aniline blue, (certified), 0.3%, phosphotungstic acid, 0.8% and acetic acid, 1.0%. Reagent stains collagen blue
  • For in vitro diagnostic use
Specifications
Specifications
Dark Purple
Safety and Handling
Safety and Handling
ShelfLife : Expiry date on the label