Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Trichrome Stain (Masson) Kit

Intended for use in the study of connective tissue, muscle and collagen fibers

Manufacturer:  Sigma Diagnostics™ HT151KT

Catalog No. SDHT15


Includes:
  • Biebrich Scarlet-Acid Fuchsin Solution (HT15-1), 250mL
  • Phosphomolybdic Acid Solution (HT15-3), 250mL
  • Phosphotungstic Acid Solution (HT15-2), 250mL
  • Aniline Blue Solution (kit only), 250mL

Description

Description

  • Collagen and muscle fiber stain differentially when treated sequentially with Biebrich Scarlet-Acid Fuchsin, Phosphotungstic/Phosphomolybdic Acid, and Aniline Blue
  • Cytoplasm and muscle fibers stain red; collagen displays blue coloration

For in vitro diagnostic use

Specifications

Specifications

Study of diseases of connective tissue and muscle characterized by fibrotic and dystrophic changes, in vitro diagnostic use
Trichrome Stain (Masson) Kit
Store reagents at room temperature (18–26°C)
  • Biebrich Scarlet-Acid Fuchsin Solution (HT15-1), 250mL
  • Phosphomolybdic Acid Solution (HT15-3), 250mL
  • Phosphotungstic Acid Solution (HT15-2), 250mL
  • Aniline Blue Solution (kit only), 250mL
1 kit
Safety and Handling

Safety and Handling

ShelfLife : Expiry date on the label

SDS
Documents
Product Certifications
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit