Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Neutral Red Solution

Used as red nuclear counterstain in various cytochemical procedures

Manufacturer:  Sigma Diagnostics™ N626450ML

Catalog No. SDN6264


Add to cart

Description

Description

  • Concentration: 0.5% (w/v, acetate buffer)
  • With added preservative

Specifications

Specifications

50mL
Solution
553-24-2
288.78g/mol
Neutral Red solution
Dark Red-Brown
As a red nuclear counterstain in various cytochemical procedures
C15H17ClN4
3-Amino-7-dimethylamino-2-methylphenazine hydrochloride, Toluylene red
Safety and Handling

Safety and Handling

ShelfLife : Expiry date on the label

SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For in vitro diagnostic use