Learn More
Sigma Diagnostics™ Fast Green FCF Dye
Used in protein staining in IEF, native PAGE and SDS-PAGE
Manufacturer:  Sigma Diagnostics™ F725825G
Catalog No. SDF7258
Add to cart
Description
Description
  • Certified for safranin-fast green FCF stain for plant cells
  • As substitute for aniline blue in Gomori's one-step trichrome procedure
Certified by the Biological Stain Commission
Specifications
Specifications
25g
1mg/mL
Red to Red-Brown
2353-45-9
808.85g/mol
Fast Green FCF
Powder
Certified for the Safranin fast green FCF Stain for plant cells, as a substitute for Aniline blue in gomori's one step trichrome procedure
290°C
C37H34N2Na2O10S3
Food green 3