Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Pseudo™ Corpse Scent (Formulation I), MilliporeSigma™ Supelco™
$116.00 - $329.00
Specifications
Type | Standard |
---|---|
Product Line | Sigma Pseudo |
MDL Number | MFCD00165840 |
Physical Form | Neat |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-100-4737
|
MilliporeSigma Supelco
P43044X1ML |
4 x 1 mL |
Each for $116.00
|
|
|||||
11-100-4399
|
MilliporeSigma Supelco
P43048X1ML |
8 x 1 mL |
Each for $218.00
|
|
|||||
11-100-4118
|
MilliporeSigma Supelco
P430420ML |
20 mL |
Each for $329.00
|
|
|||||
Specifications
Standard | |
Sigma Pseudo | |
MFCD00165840 | |
Neat |
Safety and Handling
Recommended Storage : 2°C to 8°C
Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title