Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Pseudo™ Narcotic Scent (Cocaine Formulation Powder), MilliporeSigma™ Supelco™

$59.60 - $554.50

Specifications

Product Line Sigma Pseudo
Physical Form Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-4977
View Documents
MilliporeSigma Supelco
P2423100G
100 g
Each for $59.60
Add to cart
 
11-100-3598
View Documents
MilliporeSigma Supelco
P24231KG
1 kg
Each for $554.50
Add to cart
 
Description

Description

Specifications

Specifications

Sigma Pseudo
Powder
Safety and Handling

Safety and Handling

Recommended Storage : -20°C

SDS
Product Certifications

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit