Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Pseudo™ Narcotic Scent (Heroin Formulation Powder), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
100 g
1 kg
Specifications
Specifications
| CAS | 50-78-2 |
| Product Line | Sigma Pseudo |
| Physical Form | Powder |
| MDL Number | MFCD00165842 |
| Type | Standard |
| Quantity | 1 kg |
Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction