Sigma-Transwab Liquid Amies Media Swabs with Swab Capture

Liquid transport for automated processing systems and conventional inoculation and plating

Manufacturer: ADVANTAGE BUNDLING SP LLC MW177S

 View more versions of this product

Catalog No. 23-691-020

  Check Availability
Add to Cart

    Specifications

    Bottom Shape Conical
    Bud Material Polyurethane
    Certifications/Compliance CE, M-40
    Closure Type Screw cap
    Color Orange
    For Use With (Application) For aerobic, anaerobic and fastidious microorganisms
    Includes Liquid amies medium
    Product Type MiniTip Swab

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit